missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorophenol 98.0+%, TCI America™

Catalog No. T16161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16161G 1 g
T16165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16161G Supplier TCI America Supplier No. T16161G
Only null left

Chemical Identifiers

CAS 2822-41-5
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.084
MDL Number MFCD00042426
InChI Key IJGSULQFKYOYEU-UHFFFAOYSA-N
PubChem CID 123174
IUPAC Name 2,3,4-trifluorophenol
SMILES C1=CC(=C(C(=C1O)F)F)F

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1325
Synonym phenol, 2,3,4-trifluoro, trifluorophenol, phenol,2,3,4-trifluoro, qr bf cf df, pubchem4156, acmc-209h1g, ksc497k2f, 2,3,4-tris fluoranyl phenol, 2,3,4-trifluorophenol, ijgsulqfkyoyeu-uhfffaoysa
Formula Weight 148.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trifluorophenol