missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzoic Acid 98.0+%, TCI America™

Catalog No. T18671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18671G Supplier TCI America Supplier No. T18671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61079-72-9
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.09
MDL Number MFCD00061232
InChI Key WEPXLRANFJEOFZ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzoicacid, pubchem1345, acmc-209mos, ksc354m6t, 2,3 4-trifluorobenzoic acid, 2,3,4-trifluoro-benzoic acid, rarechem al bo 0257, timtec-bb sbb006557, attercop-chm at111721, buttpark 30\01-45
PubChem CID 302932
IUPAC Name 2,3,4-trifluorobenzoic acid
SMILES OC(=O)C1=CC=C(F)C(F)=C1F

Specifications

Melting Point 141°C
Color White
Quantity 1 g
Formula Weight 176.09
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trifluorobenzoic Acid