accessibility menu, dialog, popup

UserName

CAS RN 61079-72-9

IUPAC Name: 2,3,4-trifluorobenzoic acid
Synonyms: 2,3,4-trifluorobenzoic acid
Molecular Weight (g/mol): 176.09
Molecular Formula: C7H3F3O2
InChi Key: WEPXLRANFJEOFZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results