missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trifluorobenzoic acid, 98%
CAS: 61079-72-9 | C7H3F3O2 | 176.09 g/mol
$187.12 - $707.89
Chemical Identifiers
| CAS | 61079-72-9 |
|---|---|
| Molecular Formula | C7H3F3O2 |
| Molecular Weight (g/mol) | 176.09 |
| MDL Number | MFCD00061232 |
| InChI Key | WEPXLRANFJEOFZ-UHFFFAOYSA-N |
| Synonym | 2,3,4-trifluorobenzoicacid, pubchem1345, acmc-209mos, ksc354m6t, 2,3 4-trifluorobenzoic acid, 2,3,4-trifluoro-benzoic acid, rarechem al bo 0257, timtec-bb sbb006557, attercop-chm at111721, buttpark 30\01-45 |
| PubChem CID | 302932 |
| IUPAC Name | 2,3,4-trifluorobenzoic acid |
| SMILES | OC(=O)C1=CC=C(F)C(F)=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2069606
|
Thermo Scientific Chemicals
B2069606 |
5 g |
Each for $187.12
|
|
|||||
|
AAB2069614
|
Thermo Scientific Chemicals
B2069614 |
25 g |
Each for $707.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 61079-72-9 | |
| 176.09 | |
| WEPXLRANFJEOFZ-UHFFFAOYSA-N | |
| 302932 | |
| OC(=O)C1=CC=C(F)C(F)=C1F |
| C7H3F3O2 | |
| MFCD00061232 | |
| 2,3,4-trifluorobenzoicacid, pubchem1345, acmc-209mos, ksc354m6t, 2,3 4-trifluorobenzoic acid, 2,3,4-trifluoro-benzoic acid, rarechem al bo 0257, timtec-bb sbb006557, attercop-chm at111721, buttpark 30\01-45 | |
| 2,3,4-trifluorobenzoic acid |
Specifications
| 61079-72-9 | |
| C7H3F3O2 | |
| 5 g | |
| 2,3,4-trifluorobenzoicacid, pubchem1345, acmc-209mos, ksc354m6t, 2,3 4-trifluorobenzoic acid, 2,3,4-trifluoro-benzoic acid, rarechem al bo 0257, timtec-bb sbb006557, attercop-chm at111721, buttpark 30\01-45 | |
| OC(=O)C1=CC=C(F)C(F)=C1F | |
| 176.09 | |
| 176.1 | |
| 2,3,4-Trifluorobenzoic acid |
| 140°C to 142°C | |
| MFCD00061232 | |
| 7476020 | |
| WEPXLRANFJEOFZ-UHFFFAOYSA-N | |
| 2,3,4-trifluorobenzoic acid | |
| 302932 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
RUO – Research Use Only