missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzoic acid, 98%

Catalog No. AAB2069606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2069606 5 g
AAB2069614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2069606 Supplier Thermo Scientific Chemicals Supplier No. B2069606
Only null left

CAS: 61079-72-9 | C7H3F3O2 | 176.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61079-72-9
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.09
MDL Number MFCD00061232
InChI Key WEPXLRANFJEOFZ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzoicacid, pubchem1345, acmc-209mos, ksc354m6t, 2,3 4-trifluorobenzoic acid, 2,3,4-trifluoro-benzoic acid, rarechem al bo 0257, timtec-bb sbb006557, attercop-chm at111721, buttpark 30\01-45
PubChem CID 302932
IUPAC Name 2,3,4-trifluorobenzoic acid
SMILES OC(=O)C1=CC=C(F)C(F)=C1F

Specifications

Melting Point 140°C to 142°C
Quantity 5 g
Beilstein 7476020
Formula Weight 176.1
Percent Purity 98%
Chemical Name or Material 2,3,4-Trifluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes

RUO – Research Use Only