missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trichloronitrobenzene 99.0+%, TCI America™

Catalog No. T119725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T119725G 25 g
T1197500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T119725G Supplier TCI America Supplier No. T119725G
Only null left

Chemical Identifiers

CAS 17700-09-3
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00007063
InChI Key BGKIECJVXXHLDP-UHFFFAOYSA-N
Synonym 2,3,4-trichloronitrobenzene, trichloronitrobenzene, 2,3,4-trichloro-1-nitrobenzene, 4-nitro-1,2,3-trichlorobenzene, benzene, 1,2,3-trichloro-4-nitro, benzene, 4-nitro-1,2,3-trichloro, 1,2,3-trichloronitrobenzene, 1,2,3-tris chloranyl-4-nitro-benzene, pubchem8437, acmc-1bpdn
PubChem CID 28697
IUPAC Name 1,2,3-trichloro-4-nitrobenzene
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])Cl)Cl)Cl

Specifications

Melting Point 56°C
Quantity 25 g
Formula Weight 226.44
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trichloronitrobenzene
RTECSNumber DA6612000
TSCA No