missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dihydroxyphenyl)ethyl Alcohol 98.0+%, TCI America™

Catalog No. d27561g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27561G 1 g
D27565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27561G Supplier TCI America Supplier No. D27561G
Only null left

Chemical Identifiers

CAS 10597-60-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD01320529
InChI Key JUUBCHWRXWPFFH-UHFFFAOYSA-N
Synonym 3,4-Dihydroxyphenethyl Alcohol, 2-(3,4-Dihydroxyphenyl)ethanol
PubChem CID 82755
ChEBI CHEBI:68889
IUPAC Name 4-(2-hydroxyethyl)benzene-1,2-diol
SMILES C1=CC(=C(C=C1CCO)O)O

Specifications

Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3,4-Dihydroxyphenyl)ethyl Alcohol