accessibility menu, dialog, popup

UserName

CAS RN 10597-60-1

IUPAC Name: 4-(2-hydroxyethyl)benzene-1,2-diol
Synonyms: hydroxytyrosol dopet
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: JUUBCHWRXWPFFH-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)