missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,6-Tetra-O-benzyl-D-glucopyranose 97.0+%, TCI America™
$58.85 - $159.49
Chemical Identifiers
| CAS | 4132-28-9 |
|---|---|
| Molecular Formula | C34H36O6 |
| Molecular Weight (g/mol) | 540.656 |
| MDL Number | MFCD00066004 |
| InChI Key | OGOMAWHSXRDAKZ-BKJHVTENSA-N |
| Synonym | 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose |
| PubChem CID | 10940502 |
| IUPAC Name | (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol |
| SMILES | C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5 |
Chemical Identifiers
| 4132-28-9 | |
| 540.656 | |
| OGOMAWHSXRDAKZ-BKJHVTENSA-N | |
| 10940502 | |
| C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5 |
| C34H36O6 | |
| MFCD00066004 | |
| 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose | |
| (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol |
Specifications
| 4132-28-9 | |
| White | |
| MFCD00066004 | |
| OGOMAWHSXRDAKZ-BKJHVTENSA-N | |
| (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol | |
| 10940502 | |
| ≥97.0% (HPLC) | |
| 2,3,4,6-Tetra-O-benzyl-D-glucopyranose |
| 150°C | |
| C34H36O6 | |
| 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose | |
| C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5 | |
| 540.656 | |
| 540.66 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator