missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzyl-D-glucopyranose 97.0+%, TCI America™

Catalog No. T19141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19141G 1 g
T19145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19141G Supplier TCI America Supplier No. T19141G

Chemical Identifiers

CAS 4132-28-9
Molecular Formula C34H36O6
Molecular Weight (g/mol) 540.656
MDL Number MFCD00066004
InChI Key OGOMAWHSXRDAKZ-BKJHVTENSA-N
Synonym 2,3,4,6-tetra-o-benzyl-d-glucopyranose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, tetrabenzylglucose, 3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, d-glucopyranose, 2,3,4,6-tetrakis-o-phenylmethyl, ksc235o0p, 2,3,4,6-tetra-o-benzylglucopyranose, 2,3,4,6-tetra-o-benzyl glucopyranose, 2,3,4,6-tetra-o-benzyl-glucopyranose, 2,3,4,6-terta-o-benzyl-d-glucopyranose
PubChem CID 10940502
IUPAC Name (3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 150°C
Color White
Quantity 1 g
Formula Weight 540.66
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
TSCA No
Recommended Storage Refrigerator