missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose, 98%

Catalog No. p-7030748
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3159503 1 g
AAH3159506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3159503 Supplier Thermo Scientific Chemicals Supplier No. H3159503
Only null left

CAS: 6564-72-3 | C34H36O6 | 540.656 g/mol

It is the intermediate of Voglibose/Dapagliflozin. An important D-glucopyranose derivative for glucosylations and other reactions 1,2; Preparation of the α-glucopyranosyl chloride, synthesis of 1-C-α-D-glucopyranose derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate of Voglibose/Dapagliflozin. An important D-glucopyranose derivative for glucosylations and other reactions 1,2; Preparation of the α-glucopyranosyl chloride, synthesis of 1-C-α-D-glucopyranose derivatives.

Solubility
Soluble in CHCl3. Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 6564-72-3
Molecular Formula C34H36O6
Molecular Weight (g/mol) 540.656
MDL Number MFCD00023849
InChI Key OGOMAWHSXRDAKZ-RUOAZZEASA-N
Synonym 2s,3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol, 2,3,4,6-tetra-o-benzyl-a-d-glucopyranose, 2,3,4,6-tetra-o-benzyl-alpha-glucopyranose, 2,3,4,6-tetra-o-benzyl-alpha-d-glucopyranose, 2s,3r,4s,5r,6r-3,4,5-tris benzyloxy-6-benzyloxy methyl oxan-2-ol, pubchem21934, pubchem24349, voglibose tbg impurity, 2,3,4,6-tetra-o-benzyl-alpha-d-glucose, 2,3,4,6-tetra-o-benzyl-alpha-d-gluco-pyranose
PubChem CID 11731256
IUPAC Name (2S,3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 150°C to 151°C
Quantity 1 g
Solubility Information Soluble in CHCl3. Insoluble in water.
Formula Weight 540.65
Percent Purity 98%
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only