accessibility menu, dialog, popup

UserName

OOOHOOO

CAS RN 6564-72-3

IUPAC Name: 3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Synonyms: 3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular Weight (g/mol): 540.66
Molecular Formula: C34H36O6
InChi Key: OGOMAWHSXRDAKZ-UHFFFAOYNA-N
SMILES: OC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results