missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluoroaniline 98.0+%, TCI America™

Catalog No. p-7137415
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T27821G 1 g
T27825G 5 g
2 options

Catalog No. T27821G

Supplier: TCI America T27821G

Only null left

Chemical Identifiers

CAS 5580-80-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD00025153
InChI Key BEECAQIHCYTZHC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro-phenylamine, benzenamine, 2,3,4,5-tetrafluoro, aniline, 2,3,4,5-tetrafluoro, benzenamine, tetrafluoro, aniline, 2,3,4,5-tetrafluoro-8ci, pubchem1547, acmc-1ajyg, 2,4,5-tetrafluoroaniline, ksc273e5j, aniline,3,4,5-tetrafluoro
PubChem CID 21757
IUPAC Name 2,3,4,5-tetrafluoroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)F)N

Specifications

Melting Point 28°C
Color White
Boiling Point 78°C
Quantity 1 g
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,4,5-Tetrafluoroaniline