accessibility menu, dialog, popup

UserName

CAS RN 5580-80-3

IUPAC Name: 2,3,4,5-tetrafluoroaniline
Synonyms: 2,3,4,5-tetrafluoroaniline
Molecular Weight (g/mol): 165.09
Molecular Formula: C6H3F4N
InChi Key: BEECAQIHCYTZHC-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)