missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluoroaniline 98.0+%, TCI America™

Catalog No. T27825G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27825G 5 g
T27821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27825G Supplier TCI America Supplier No. T27825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5580-80-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD00025153
InChI Key BEECAQIHCYTZHC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro-phenylamine, benzenamine, 2,3,4,5-tetrafluoro, aniline, 2,3,4,5-tetrafluoro, benzenamine, tetrafluoro, aniline, 2,3,4,5-tetrafluoro-8ci, pubchem1547, acmc-1ajyg, 2,4,5-tetrafluoroaniline, ksc273e5j, aniline,3,4,5-tetrafluoro
PubChem CID 21757
IUPAC Name 2,3,4,5-tetrafluoroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)F)N

Specifications

Melting Point 28°C
Color White
Boiling Point 78°C
Quantity 5 g
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,4,5-Tetrafluoroaniline