missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5-Tetrafluoroaniline, 98%

Catalog No. AAH6141703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6141703 1 g
AAH6141706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6141703 Supplier Thermo Scientific Chemicals Supplier No. H6141703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5580-80-3 | C6H3F4N | 165.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5580-80-3
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD00025153
InChI Key BEECAQIHCYTZHC-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro-phenylamine, benzenamine, 2,3,4,5-tetrafluoro, aniline, 2,3,4,5-tetrafluoro, benzenamine, tetrafluoro, aniline, 2,3,4,5-tetrafluoro-8ci, pubchem1547, acmc-1ajyg, 2,4,5-tetrafluoroaniline, ksc273e5j, aniline,3,4,5-tetrafluoro
PubChem CID 21757
IUPAC Name 2,3,4,5-tetrafluoroaniline
SMILES C1=C(C(=C(C(=C1F)F)F)F)N

Specifications

Melting Point 27°C to 29°C
Color White to Brown
Boiling Point 78°C
Flash Point 79°C (174°F)
Quantity 1 g
UN Number UN2811
Formula Weight 165.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,4,5-Tetrafluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only