missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,5,6-Pentafluorotoluene 98.0+%, TCI America™
$95.06 - $462.29
Chemical Identifiers
| CAS | 771-56-2 |
|---|---|
| Molecular Formula | C7H3F5 |
| Molecular Weight (g/mol) | 182.093 |
| MDL Number | MFCD00000298 |
| InChI Key | SXPRVMIZFRCAGC-UHFFFAOYSA-N |
| Synonym | 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene |
| PubChem CID | 69869 |
| IUPAC Name | 1,2,3,4,5-pentafluoro-6-methylbenzene |
| SMILES | CC1=C(C(=C(C(=C1F)F)F)F)F |
Chemical Identifiers
| 771-56-2 | |
| 182.093 | |
| SXPRVMIZFRCAGC-UHFFFAOYSA-N | |
| 69869 | |
| CC1=C(C(=C(C(=C1F)F)F)F)F |
| C7H3F5 | |
| MFCD00000298 | |
| 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene | |
| 1,2,3,4,5-pentafluoro-6-methylbenzene |
Specifications
| 771-56-2 | |
| C7H3F5 | |
| 5 g | |
| 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene | |
| CC1=C(C(=C(C(=C1F)F)F)F)F | |
| 182.093 | |
| 182.09 | |
| Liquid |
| Colorless | |
| MFCD00000298 | |
| 1993 | |
| SXPRVMIZFRCAGC-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentafluoro-6-methylbenzene | |
| 69869 | |
| ≥98.0% (GC) | |
| 2,3,4,5,6-Pentafluorotoluene |
Safety and Handling
TSCA : Yes