CAS RN 771-56-2
IUPAC Name:
1,2,3,4,5-pentafluoro-6-methylbenzene
Synonyms:
N-methylpentafluorobenzene
Molecular Weight (g/mol):
182.09
Molecular Formula:
C7H3F5
InChi Key:
SXPRVMIZFRCAGC-UHFFFAOYSA-N
SMILES:
CC1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
of
1
results
1
2,3,4,5,6-Pentafluorotoluene 98.0+%, TCI America™
CAS: 771-56-2 Molecular Formula: C7H3F5 Molecular Weight (g/mol): 182.093 MDL Number: MFCD00000298 InChI Key: SXPRVMIZFRCAGC-UHFFFAOYSA-N Synonym: 2,3,4,5,6-pentafluorotoluene,pentafluorotoluene,methyl pentafluorobenzene,pentafluoromethylbenzene,benzene, pentafluoromethyl,methylpentafluorobenzene,benzene, 1,2,3,4,5-pentafluoro-6-methyl,toluene, 2,3,4,5,6-pentafluoro,2,3,4,5,6-pentafluoro-toluene,n-methylpentafluorobenzene PubChem CID: 69869 IUPAC Name: 1,2,3,4,5-pentafluoro-6-methylbenzene SMILES: CC1=C(C(=C(C(=C1F)F)F)F)F