missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorotoluene 98.0+%, TCI America™

Catalog No. P14085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14085G 5 g
P140825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14085G Supplier TCI America Supplier No. P14085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 771-56-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.093
MDL Number MFCD00000298
InChI Key SXPRVMIZFRCAGC-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene
PubChem CID 69869
IUPAC Name 1,2,3,4,5-pentafluoro-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 182.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorotoluene