missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorostyrene, 97%, stabilized

CAS: 653-34-9 | C8H3F5 | 194.1 g/mol

$132.69 - $493.94

Chemical Identifiers

CAS 653-34-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.1
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC188470050
SDS View Documents
Thermo Scientific Chemicals
188470050
5 g Glass bottle
Each for $132.69
Only null left
 
AC188470250
SDS View Documents
Thermo Scientific Chemicals
188470250
25 g Glass bottle
Each for $493.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

653-34-9
194.1
LVJZCPNIJXVIAT-UHFFFAOYSA-N
69556
C=CC1=C(C(=C(C(=C1F)F)F)F)F
C8H3F5
MFCD00000300
2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
1-ethenyl-2,3,4,5,6-pentafluorobenzene

Specifications

653-34-9,123-31-9
100.0
1.4060g/mL
34°C
96% min. (GC)
C8H3F5
C6F5CH=CH2
5 g
2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
C=CC1=C(C(=C(C(=C1F)F)F)F)F
194.1
69556
97%
2,3,4,5,6-Pentafluorostyrene, 97%
96.0
Colorless to Yellow
62.0°C to 63.0°C (50.0 mmHg)
Authentic
Glass bottle
1.4445 to 1.4465
MFCD00000300
1.406
LVJZCPNIJXVIAT-UHFFFAOYSA-N
1-ethenyl-2,3,4,5,6-pentafluorobenzene
0.01% tert.-butylcatechol
194.1
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai

GHS Signal Word: Warning

EINECSNumber : 211-500-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only