missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzyl alcohol, 98%

Catalog No. AAA1211906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1211906 5 g
AAA1211914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1211906 Supplier Thermo Scientific Chemicals Supplier No. A1211906

CAS: 440-60-8 | C7H3F5O | 198.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 440-60-8
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00004602
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
PubChem CID 9923
ChEBI CHEBI:44903
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O

Specifications

Melting Point 32°C to 36°C
Boiling Point 182°C
Flash Point 87°C (188°F)
Quantity 5 g
Beilstein 2052669
Formula Weight 198.09
Percent Purity 98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 207-126-7
RTECSNumber DP0695000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only