accessibility menu, dialog, popup

UserName

HOFFFFF

CAS RN 440-60-8

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)methanol
Synonyms: pentafluorobenzyl alcohol (hydroxymethyl)pentafluorobenzene
Molecular Weight (g/mol): 198.09
Molecular Formula: C7H3F5O
InChi Key: PGJYYCIOYBZTPU-UHFFFAOYSA-N
SMILES: OCC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)