missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentabromotoluene 98.0+%, TCI America™

Catalog No. p-7134795
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P082725G Supplier TCI America Supplier No. P082725G

Chemical Identifiers

CAS 87-83-2
Molecular Formula C7H3Br5
Molecular Weight (g/mol) 486.621
MDL Number MFCD00000060
InChI Key OZHJEQVYCBTHJT-UHFFFAOYSA-N
PubChem CID 6906
IUPAC Name 1,2,3,4,5-pentabromo-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1Br)Br)Br)Br)Br

Specifications

Melting Point 290°C
Quantity 25 g
UN Number 3077
Formula Weight 486.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5,6-Pentabromotoluene
EINECSNumber (3)-0058
RTECSNumber DA6639500
TSCA Yes