accessibility menu, dialog, popup

UserName

CH3BrBrBrBrBr

CAS RN 87-83-2

IUPAC Name: 1,2,3,4,5-pentabromo-6-methylbenzene
Synonyms: pentabromotoluene
Molecular Weight (g/mol): 486.62
Molecular Formula: C7H3Br5
InChi Key: OZHJEQVYCBTHJT-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(Br)=C(Br)C(Br)=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results