missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4,4'-Tetrahydroxydiphenylmethane 95.0+%, TCI America™
Supplier: TCI America T170725G
Specifications
| 2,3,4,4′-Tetrahydroxydiphenylmethane | |
| 151°C | |
| MFCD01321194 | |
| NBTCXIZQSZQNKJ-UHFFFAOYSA-N | |
| 4-[(4-hydroxyphenyl)methyl]benzene-1,2,3-triol | |
| 21896295 | |
| ≥95.0% (GC) |
| 174462-43-2 | |
| C13H12O4 | |
| 25 g | |
| OC1=CC=C(CC2=C(O)C(O)=C(O)C=C2)C=C1 | |
| 232.24 | |
| 232.24 | |
| Crystalline Powder |
Chemical Identifiers
| 174462-43-2 | |
| 232.24 | |
| NBTCXIZQSZQNKJ-UHFFFAOYSA-N | |
| 4-[(4-hydroxyphenyl)methyl]benzene-1,2,3-triol |
| C13H12O4 | |
| MFCD01321194 | |
| 21896295 | |
| OC1=CC=C(CC2=C(O)C(O)=C(O)C=C2)C=C1 |
Safety and Handling
TSCA : No