accessibility menu, dialog, popup

UserName

OHOHHOOH

CAS RN 174462-43-2

IUPAC Name: 4-[(4-hydroxyphenyl)methyl]benzene-1,2,3-triol
Synonyms: 4-[(4-hydroxyphenyl)methyl]benzene-1,2,3-triol
Molecular Weight (g/mol): 232.24
Molecular Formula: C13H12O4
InChi Key: NBTCXIZQSZQNKJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC2=C(O)C(O)=C(O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results