missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,4'-Tetrahydroxydiphenylmethane 95.0+%, TCI America™

Catalog No. T17075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17075G 5 g
T170725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17075G Supplier TCI America Supplier No. T17075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174462-43-2
Molecular Formula C13H12O4
Molecular Weight (g/mol) 232.24
MDL Number MFCD01321194
InChI Key NBTCXIZQSZQNKJ-UHFFFAOYSA-N
PubChem CID 21896295
IUPAC Name 4-[(4-hydroxyphenyl)methyl]benzene-1,2,3-triol
SMILES OC1=CC=C(CC2=C(O)C(O)=C(O)C=C2)C=C1

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 232.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,4′-Tetrahydroxydiphenylmethane