missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol 98.0+%, TCI America™
$52.19 - $231.14
Chemical Identifiers
| CAS | 70321-86-7 |
|---|---|
| Molecular Formula | C30H29N3O |
| Molecular Weight (g/mol) | 447.582 |
| MDL Number | MFCD00134705 |
| InChI Key | OLFNXLXEGXRUOI-UHFFFAOYSA-N |
| PubChem CID | 112412 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol |
| SMILES | CC(C)(C1=CC=CC=C1)C2=CC(=C(C(=C2)N3N=C4C=CC=CC4=N3)O)C(C)(C)C5=CC=CC=C5 |
Chemical Identifiers
| 70321-86-7 | |
| 447.582 | |
| OLFNXLXEGXRUOI-UHFFFAOYSA-N | |
| 2-(benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol |
| C30H29N3O | |
| MFCD00134705 | |
| 112412 | |
| CC(C)(C1=CC=CC=C1)C2=CC(=C(C(=C2)N3N=C4C=CC=CC4=N3)O)C(C)(C)C5=CC=CC=C5 |
Specifications
| 70321-86-7 | |
| White-Yellow | |
| MFCD00134705 | |
| OLFNXLXEGXRUOI-UHFFFAOYSA-N | |
| 2-(benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol | |
| 112412 | |
| ≥98.0% (HPLC) | |
| 2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol |
| 140°C | |
| C30H29N3O | |
| 25 g | |
| CC(C)(C1=CC=CC=C1)C2=CC(=C(C(=C2)N3N=C4C=CC=CC4=N3)O)C(C)(C)C5=CC=CC=C5 | |
| 447.582 | |
| 447.58 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-5459
TSCA : Yes