accessibility menu, dialog, popup

UserName

CAS RN 70321-86-7

H3CCH3NNNOHCH3H3C

CAS RN 70321-86-7

IUPAC Name: 2-(2H-1,2,3-benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol
Synonyms: 2-(1,2,3-benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol
Molecular Weight (g/mol): 447.58
Molecular Formula: C30H29N3O
InChi Key: OLFNXLXEGXRUOI-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC=CC=C1)C1=CC(N2N=C3C=CC=CC3=N2)=C(O)C(=C1)C(C)(C)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results