Learn More
2,2'-Thiodiethanethiol, tech. 85%
CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol
Supplier: Thermo Scientific Chemicals B2230418
Description
2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2,2'-Thiodiethanethiol | |
| 1.183 | |
| 90°C (194°F) | |
| 1.5955 | |
| 50 g | |
| Difficult to mix. | |
| SCCSCCS | |
| 154.30 | |
| 154.32 |
| 3570-55-6 | |
| 135°C to 136°C (10 mmHg) | |
| C4H10S3 | |
| MFCD00004893 | |
| 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi | |
| KSJBMDCFYZKAFH-UHFFFAOYSA-N | |
| 2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol | |
| 77117 | |
| ≥90% |
Chemical Identifiers
| 3570-55-6 | |
| 154.30 | |
| KSJBMDCFYZKAFH-UHFFFAOYSA-N | |
| 77117 |
| C4H10S3 | |
| MFCD00004893 | |
| 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi | |
| SCCSCCS |
Safety and Handling
GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 222-671-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only