missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Phenylsulfonylmethyl)phenyl]-1,3-dioxolane 97.0+%, TCI America™

Catalog No. p1454100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1454100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1454100MG Supplier TCI America Supplier No. P1454100MG

Chemical Identifiers

CAS 226089-80-1
Molecular Formula C16H16O4S
Molecular Weight (g/mol) 304.36
MDL Number MFCD06797156
InChI Key ANNWDOBNPSVXNI-UHFFFAOYSA-N
Synonym 2-[2-(Benzenesulfonylmethyl)phenyl]-1,3-dioxolane, 2-(1,3-Dioxolan-2-yl)benzyl Phenyl Sulfone
PubChem CID 44630458
IUPAC Name 2-[2-(benzenesulfonylmethyl)phenyl]-1,3-dioxolane
SMILES C1COC(O1)C2=CC=CC=C2CS(=O)(=O)C3=CC=CC=C3

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 304.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-(Phenylsulfonylmethyl)phenyl]-1,3-dioxolane