accessibility menu, dialog, popup

UserName

OOSOO

CAS RN 226089-80-1

IUPAC Name: 2-{2-[(benzenesulfonyl)methyl]phenyl}-1,3-dioxolane
Synonyms: 2-{2-[(benzenesulfonyl)methyl]phenyl}-1,3-dioxolane
Molecular Weight (g/mol): 304.36
Molecular Formula: C16H16O4S
InChi Key: ANNWDOBNPSVXNI-UHFFFAOYSA-N
SMILES: O=S(=O)(CC1=CC=CC=C1C1OCCO1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results