missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
CAS: 104206-65-7 | C14H10F3NO5 | 329.231 g/mol
$194.18 - $577.26
Chemical Identifiers
| CAS | 104206-65-7 |
|---|---|
| Molecular Formula | C14H10F3NO5 |
| Molecular Weight (g/mol) | 329.231 |
| MDL Number | MFCD01752192 |
| InChI Key | OUBCNLGXQFSTLU-UHFFFAOYSA-N |
| Synonym | nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona |
| PubChem CID | 115355 |
| ChEBI | CHEBI:50378 |
| IUPAC Name | 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione |
| SMILES | C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH66633MD
|
Thermo Scientific Chemicals
H66633MD |
250 mg |
Each for $194.18
|
|
|||||
|
AAH6663303
|
Thermo Scientific Chemicals
H6663303 |
1 g |
Each for $577.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104206-65-7 | |
| 329.231 | |
| OUBCNLGXQFSTLU-UHFFFAOYSA-N | |
| 115355 | |
| 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione |
| C14H10F3NO5 | |
| MFCD01752192 | |
| nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona | |
| CHEBI:50378 | |
| C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-] |
Specifications
| 104206-65-7 | |
| MFCD01752192 | |
| nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona | |
| C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-] | |
| 329.231 | |
| CHEBI:50378 | |
| 95% | |
| 2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione |
| C14H10F3NO5 | |
| 250 mg | |
| OUBCNLGXQFSTLU-UHFFFAOYSA-N | |
| 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione | |
| 115355 | |
| 329.23 | |
| Solid |
Safety and Handling
RTECSNumber : GV0766666
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only