missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%

CAS: 104206-65-7 | C14H10F3NO5 | 329.231 g/mol

Supplier:  Thermo Scientific Chemicals H6663303

 View more versions of this product

Catalog No. AAH6663303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione
C14H10F3NO5
1 g
OUBCNLGXQFSTLU-UHFFFAOYSA-N
2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
115355
329.23
Solid
104206-65-7
MFCD01752192
nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]
329.231
CHEBI:50378
95%

Chemical Identifiers

104206-65-7
329.231
OUBCNLGXQFSTLU-UHFFFAOYSA-N
115355
2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
C14H10F3NO5
MFCD01752192
nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
CHEBI:50378
C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GV0766666

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only