missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) 98.0+%, TCI America™

Catalog No. p-7132060
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021525G 25 g
M0215500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M021525G Supplier TCI America Supplier No. M021525G

Chemical Identifiers

CAS 88-24-4
Molecular Formula C25H36O2
Molecular Weight (g/mol) 368.561
MDL Number MFCD00026296
InChI Key GPNYZBKIGXGYNU-UHFFFAOYSA-N
PubChem CID 6928
IUPAC Name 2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
SMILES CCC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)CC)C(C)(C)C)O

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Synonym Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, 3,3′C-Di-tert-butyl-5,5′C-diethyl-2,2′C-dihydroxydiphenylmethane, 2,2′C-Methylenebis(4-ethyl-6-tert-butylphenol)
Formula Weight 368.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
EINECSNumber (4)-0093
RTECSNumber SL9800000
TSCA Yes