missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Methylenebis(4-methylphenol) 90.0+%, TCI America™
$113.71 - $342.98
Chemical Identifiers
| CAS | 3236-63-3 |
|---|---|
| Molecular Formula | C15H16O2 |
| Molecular Weight (g/mol) | 228.29 |
| MDL Number | MFCD00155176 |
| InChI Key | XZXYQEHISUMZAT-UHFFFAOYSA-N |
| PubChem CID | 76715 |
| IUPAC Name | 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol |
| SMILES | CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1 |
Chemical Identifiers
| 3236-63-3 | |
| 228.29 | |
| XZXYQEHISUMZAT-UHFFFAOYSA-N | |
| 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol |
| C15H16O2 | |
| MFCD00155176 | |
| 76715 | |
| CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1 |
Specifications
| 3236-63-3 | |
| White | |
| MFCD00155176 | |
| Bis(2-hydroxy-5-methylphenyl)methane, 2,2′C-Dihydroxy-5,5′C-dimethyldiphenylmethane, 2,2′C-Methylenebis(p-cresol) | |
| CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1 | |
| 228.29 | |
| 228.29 | |
| Crystalline Powder |
| 125°C | |
| C15H16O2 | |
| 5 g | |
| XZXYQEHISUMZAT-UHFFFAOYSA-N | |
| 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol | |
| 76715 | |
| ≥90.0% (GC) | |
| 2,2′-Methylenebis(4-methylphenol) |
Safety and Handling
TSCA : Yes