missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Methylenebis(4-methylphenol) 90.0+%, TCI America™
Supplier: TCI America M113425G
Specifications
| 2,2′-Methylenebis(4-methylphenol) | |
| 125°C | |
| C15H16O2 | |
| 25 g | |
| XZXYQEHISUMZAT-UHFFFAOYSA-N | |
| 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol | |
| 76715 | |
| ≥90.0% (GC) |
| 3236-63-3 | |
| White | |
| MFCD00155176 | |
| Bis(2-hydroxy-5-methylphenyl)methane, 2,2′-Dihydroxy-5,5′-dimethyldiphenylmethane, 2,2′-Methylenebis(p-cresol) | |
| CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1 | |
| 228.29 | |
| 228.29 | |
| Crystalline Powder |
Chemical Identifiers
| 3236-63-3 | |
| 228.29 | |
| XZXYQEHISUMZAT-UHFFFAOYSA-N | |
| 2-[(2-hydroxy-5-methylphenyl)methyl]-4-methylphenol |
| C15H16O2 | |
| MFCD00155176 | |
| 76715 | |
| CC1=CC=C(O)C(CC2=CC(C)=CC=C2O)=C1 |
Safety and Handling
TSCA : Yes