missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Methylenebis(4-chlorophenol), 95%

CAS: 97-23-4 | C13H10Cl2O2 | 269.12 g/mol

Supplier:  Thermo Scientific Chemicals A1264022

 View more versions of this product

Catalog No. AAA1264022


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2'-Methylenebis(4-chlorophenol)
172°C to 176°C
MFCD00002322
UN3077
14,3071
MDNWOSOZYLHTCG-UHFFFAOYSA-N
4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
3037
269.13
97-23-4
C13H10Cl2O2
100 g
1884514
dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen
OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O
269.12
CHEBI:34689
95%

Chemical Identifiers

97-23-4
269.12
MDNWOSOZYLHTCG-UHFFFAOYSA-N
3037
4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
C13H10Cl2O2
MFCD00002322
dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen
CHEBI:34689
OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-567-1

RTECSNumber : SM0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only