missing translation for 'onlineSavingsMsg'
Learn More

(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98.0+%, TCI America™

$61.51 - $175.22

Chemical Identifiers

CAS 175166-49-1
Molecular Formula C21H18N2O2
Molecular Weight (g/mol) 330.387
MDL Number MFCD00799561
InChI Key BDHSVQLSNIGJNC-JYWFKMLOSA-N
PubChem CID 11024010
IUPAC Name (3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole
SMILES C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1402100MG
View Documents
TCI America
M1402100MG
100 mg
Each for $61.51
Only null left
 
M1402500MG
View Documents
TCI America
M1402500MG
500 mg
Each for $175.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

175166-49-1
330.387
BDHSVQLSNIGJNC-JYWFKMLOSA-N
(3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole
C21H18N2O2
MFCD00799561
11024010
C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56

Specifications

175166-49-1
White
MFCD00799561
BDHSVQLSNIGJNC-JYWFKMLOSA-N
(3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole
11024010
≥98.0% (HPLC,N)
(-)-2,2′-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
224°C
C21H18N2O2
100 mg
C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56
330.387
330.39
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'