missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98.0+%, TCI America™
Supplier: TCI America M1402500MG
Specifications
| (-)-2,2′-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] | |
| 224°C | |
| C21H18N2O2 | |
| 500 mg | |
| C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56 | |
| 330.387 | |
| 330.39 | |
| Crystalline Powder |
| 175166-49-1 | |
| White | |
| MFCD00799561 | |
| BDHSVQLSNIGJNC-JYWFKMLOSA-N | |
| (3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole | |
| 11024010 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 175166-49-1 | |
| 330.387 | |
| BDHSVQLSNIGJNC-JYWFKMLOSA-N | |
| (3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole |
| C21H18N2O2 | |
| MFCD00799561 | |
| 11024010 | |
| C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56 |
Safety and Handling
TSCA : No