missing translation for 'onlineSavingsMsg'
Learn More

(+)-2,2'-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98.0+%, TCI America™

Catalog No. M1401100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1401100MG 100 mg
M1401500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1401100MG Supplier TCI America Supplier No. M1401100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 180186-94-1
Molecular Formula C21H18N2O2
Molecular Weight (g/mol) 330.39
MDL Number MFCD06797115
InChI Key BDHSVQLSNIGJNC-UHFFFAOYNA-N
PubChem CID 1268099
IUPAC Name 2-({3aH,8H,8aH-indeno[1,2-d][1,3]oxazol-2-yl}methyl)-3aH,8H,8aH-indeno[1,2-d][1,3]oxazole
SMILES C(C1=NC2C(CC3=CC=CC=C23)O1)C1=NC2C(CC3=CC=CC=C23)O1

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 100 mg
Formula Weight 330.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (+)-2,2′-Methylenebis[(3aR,8aS)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]