missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenyl)ethylamine 98.0+%, TCI America™

Supplier:  TCI America M084425ML

 View more versions of this product

Catalog No. M084425ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(2-Methoxyphenyl)ethylamine
Yellow
C9H13NO
25 mL
2-methoxyphenethylamine, 2-2-methoxyphenyl ethylamine, 2-2-methoxyphenyl ethanamine, 2-2-methoxy-phenyl-ethylamine, 2-2-methoxyphenyl ethan-1-amine, benzeneethanamine, 2-methoxy, o-methoxyphenethylamine, pubchem23973, acmc-209fah, ortho-methoxyphenethylamine
COC1=CC=CC=C1CCN
151.209
151.21
Liquid
2045-79-6
237°C
MFCD00008186
3267
WSWPCNMLEVZGSM-UHFFFAOYSA-N
2-(2-methoxyphenyl)ethanamine
74896
≥98.0% (GC,T)

Chemical Identifiers

2045-79-6
151.209
WSWPCNMLEVZGSM-UHFFFAOYSA-N
74896
COC1=CC=CC=C1CCN
C9H13NO
MFCD00008186
2-methoxyphenethylamine, 2-2-methoxyphenyl ethylamine, 2-2-methoxyphenyl ethanamine, 2-2-methoxy-phenyl-ethylamine, 2-2-methoxyphenyl ethan-1-amine, benzeneethanamine, 2-methoxy, o-methoxyphenethylamine, pubchem23973, acmc-209fah, ortho-methoxyphenethylamine
2-(2-methoxyphenyl)ethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions