accessibility menu, dialog, popup

UserName

CH3OH3N

CAS RN 2045-79-6

IUPAC Name: 2-(2-methoxyphenyl)ethan-1-aminium
Synonyms: 2-(2-methoxyphenyl)ethanaminium
Molecular Weight (g/mol): 152.22
Molecular Formula: C9H14NO
InChi Key: WSWPCNMLEVZGSM-UHFFFAOYSA-O
SMILES: COC1=CC=CC=C1CC[NH3+]
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)