missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7132398
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08445ML 5 mL
M084425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08445ML Supplier TCI America Supplier No. M08445ML

Chemical Identifiers

CAS 2045-79-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008186
InChI Key WSWPCNMLEVZGSM-UHFFFAOYSA-N
Synonym 2-methoxyphenethylamine, 2-2-methoxyphenyl ethylamine, 2-2-methoxyphenyl ethanamine, 2-2-methoxy-phenyl-ethylamine, 2-2-methoxyphenyl ethan-1-amine, benzeneethanamine, 2-methoxy, o-methoxyphenethylamine, pubchem23973, acmc-209fah, ortho-methoxyphenethylamine
PubChem CID 74896
IUPAC Name 2-(2-methoxyphenyl)ethanamine
SMILES COC1=CC=CC=C1CCN

Specifications

Color Yellow
Boiling Point 237°C
Quantity 5 mL
UN Number 3267
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Methoxyphenyl)ethylamine