missing translation for 'onlineSavingsMsg'
Learn More

2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole 98.0+%, TCI America™

Catalog No. p-7130945
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H126125G 25 g
H1261500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H126125G Supplier TCI America Supplier No. H126125G
Only null left

Chemical Identifiers

CAS 96478-09-0
Molecular Formula C18H17N3O3
Molecular Weight (g/mol) 323.352
MDL Number MFCD00213348
InChI Key VCYCUECVHJJFIQ-UHFFFAOYSA-N
PubChem CID 726893
IUPAC Name 2-[3-(benzotriazol-2-yl)-4-hydroxyphenyl]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 25 g
Formula Weight 323.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole
EINECSNumber (5)-6574
TSCA Yes