accessibility menu, dialog, popup

UserName

CAS RN 96478-09-0

IUPAC Name: 2-[3-(2H-1,2,3-benzotriazol-2-yl)-4-hydroxyphenyl]ethyl 2-methylprop-2-enoate
Synonyms: 2-[3-(1,2,3-benzotriazol-2-yl)-4-hydroxyphenyl]ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 323.35
Molecular Formula: C18H17N3O3
InChi Key: VCYCUECVHJJFIQ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCC1=CC(N2N=C3C=CC=CC3=N2)=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results