missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

$216.24 - $644.97

Chemical Identifiers

CAS 106556-36-9
Molecular Formula C22H17N3O2
Molecular Weight (g/mol) 355.397
MDL Number MFCD23102325
InChI Key SLVHGNKODJJKEZ-UHFFFAOYSA-N
PubChem CID 53424132
IUPAC Name 6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one
SMILES COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15315G
View Documents
TCI America
H15315G
5 g
Each for $216.24
Only null left
 
H153125G
View Documents
TCI America
H153125G
25 g
Each for $644.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

106556-36-9
355.397
SLVHGNKODJJKEZ-UHFFFAOYSA-N
6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one
C22H17N3O2
MFCD23102325
53424132
COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1

Specifications

106556-36-9
Yellow
MFCD23102325
SLVHGNKODJJKEZ-UHFFFAOYSA-N
6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one
53424132
≥98.0% (HPLC,N)
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine
208°C
C22H17N3O2
5 g
COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1
355.397
355.40
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'