missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
Supplier: TCI America H153125G
Specifications
| 2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine | |
| 208°C | |
| C22H17N3O2 | |
| 25 g | |
| COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1 | |
| 355.397 | |
| 355.40 | |
| Crystalline Powder |
| 106556-36-9 | |
| Yellow | |
| MFCD23102325 | |
| SLVHGNKODJJKEZ-UHFFFAOYSA-N | |
| 6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one | |
| 53424132 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 106556-36-9 | |
| 355.397 | |
| SLVHGNKODJJKEZ-UHFFFAOYSA-N | |
| 6-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-methoxycyclohexa-2,4-dien-1-one |
| C22H17N3O2 | |
| MFCD23102325 | |
| 53424132 | |
| COC1=CC(=O)C(=C2NC(=NC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C=C1 |
Safety and Handling
TSCA : Yes