missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Fluorophenyl)ethanol 95.0+%, TCI America™
$63.52 - $250.28
Chemical Identifiers
| CAS | 50919-06-7 |
|---|---|
| Molecular Formula | C8H9FO |
| Molecular Weight (g/mol) | 140.16 |
| MDL Number | MFCD00002887 |
| InChI Key | HNIGZVZDWCTFPR-UHFFFAOYSA-N |
| Synonym | 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol |
| PubChem CID | 2733296 |
| IUPAC Name | 2-(2-fluorophenyl)ethan-1-ol |
| SMILES | OCCC1=CC=CC=C1F |
Chemical Identifiers
| 50919-06-7 | |
| 140.16 | |
| HNIGZVZDWCTFPR-UHFFFAOYSA-N | |
| 2733296 | |
| OCCC1=CC=CC=C1F |
| C8H9FO | |
| MFCD00002887 | |
| 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol | |
| 2-(2-fluorophenyl)ethan-1-ol |
Specifications
| 50919-06-7 | |
| 137°C | |
| MFCD00002887 | |
| 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol | |
| OCCC1=CC=CC=C1F | |
| 140.16 | |
| 140.16 | |
| Liquid |
| Yellow | |
| C8H9FO | |
| 5 g | |
| HNIGZVZDWCTFPR-UHFFFAOYSA-N | |
| 2-(2-fluorophenyl)ethan-1-ol | |
| 2733296 | |
| ≥95.0% (GC) | |
| 2-(2-Fluorophenyl)ethanol |
Safety and Handling
TSCA : No