accessibility menu, dialog, popup

UserName

OHF

CAS RN 50919-06-7

IUPAC Name: 2-(2-fluorophenyl)ethan-1-ol
Synonyms: 2-(2-fluorophenyl)ethanol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: HNIGZVZDWCTFPR-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC=C1F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)